A-674563 hydrochloride ≥95%

Supplier: ADIPOGEN CORP MS
Warning

SYN-1110-M001 SYN-1110-M005 SYN-1110-M010
102992-106EA 261.69 USD
102992-106 102992-108 102992-110
A-674563 hydrochloride ≥95%
A-674563 hydrochloride

A-674563 is a potent, orally available Akt1 inhibitor with an IC(50) of 11nM. It also displays activity against PKA and CDK2 with IC(50) values of 16 and 46nM respectively.

Formula: C₂₂H₂₂N₄O·HCl
Storage Temperature: Freezer
CAS Number: 552325-73-2

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR