PLX4720 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: N-(3-(5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulfonamide, PLX-4720, N-(3-(5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulphonamide

SYN-1069-M001 SYN-1069-M005 SYN-1069-M010 SYN-1069-M050
102991-792EA 90 USD
102991-792 102991-794 102991-796 102991-798
PLX4720 ≥95%
PLX4720

PLX4720 is a potent and selective inhibitor of B-Raf V600E with an IC(50) of 13nM. It is equally potent to c-Raf-1 (Y340D and Y341D mutations), and shows 10-fold selectivity for B-Raf V600E over wild-type B-Raf.

Formula: C₁₇H₁₄ClF₂N₃O₃S
Storage Temperature: Freezer
MDL Number: MFCD14635203
CAS Number: 918505-84-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR