JNJ-7706621 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: Aurora Kinase/Cdk Inhibitor, 4-((5-Amino-1-(2,6-difluorobenzoyl)-1H-1,2,4-triazol-3-yl)amino)benzenesulphonamide, 4-(5-Amino-1-(2,6-difluorobenzoyl)-1H-[1,2,4]triazol-3-ylamino)-benzenesulphonamide, Aurora A Inhibitor I, 4-(5-Amino-1-(2,6-difluorobenzoyl)-1H-[1,2,4]triazol-3-ylamino)-benzenesulfonamide, 4-((5-Amino-1-(2,6-difluorobenzoyl)-1H-1,2,4-triazol-3-yl)amino)benzenesulfonamide

SYN-1048-M001 SYN-1048-M005 SYN-1048-M010 SYN-1048-M050
102991-622EA 186.92 USD
102991-622 102991-624 102991-626 102991-628
JNJ-7706621 ≥95%
JNJ-7706621

JNJ-7706621 is a dual inhibitor of cyclin-dependent kinases (CDK) and aurora kinases. It demonstrates highest potency on CDK1/2 with IC(50) values of 9nM and 4nM respectively. It shows more than 6-fold selectivity for CDK1/2 over CDK3/4/6.

Formula: C₁₅H₁₂F₂N₆O₃S
Storage Temperature: Freezer
MDL Number: MFCD11100270
CAS Number: 443797-96-4

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR