Polyinosinic-polycytidylic acid potassium salt ≥99% endotoxin-free

Supplier: ADIPOGEN CORP MS

IAX-200-021-M002 IAX-200-021-3005 IAX-200-021-5005
102991-266EA 177.23 USD
102991-266 102991-270 102991-272
Polyinosinic-polycytidylic acid potassium salt ≥99% endotoxin-free
Polyinosinic-polycytidylic acid potassium salt

Toll-like receptor (TLR) 3 is an endosomal TLR that mediates immune responses against viral infections upon activation by its ligand double-stranded RNA, a replication intermediate of most viruses. TLR3 is expressed widely in the body and activates both the innate and adaptive immune systems. Toll-like receptor 3 (TLR3), melanoma differentiation-associated gene 5 (MDA5), and retinoic acid-inducible gene-I (RIG-I), all sensors of double-stranded RNA (dsRNA) are potent inducers of antiviral activity. dsRNA sensor activation -e.g. by poly (I:C)- induces pro-inflammatory TNF-alpha and antiviral IFN-beta, but can also enhance the expression of pro-apoptotic proteins. Recently, poly (I:C)-induced cell death recently gained considerable attention as a tool to study the 'Ripoptosome' or 'Necrosome' complex, a novel intracellular signaling complex, thought to induce regulated necrosis, also called 'Necroptosis'.

Formula: (C10H10N4NaO7P)x·(C9H11N3NaO7P)x
Storage Temperature: Refrigerator
CAS Number: 31852-29-6

Order Now

Specification Test Results

Purity 99%
Formulation Lyophilized. Sterile.
Endotoxin 0.002EU/µg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR