Zinpyr-1 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 9-(2-Carboxyphenyl)-2,7-dichloro-4,5-bis[di(2-pyridyl)aminomethyl]-6-hydroxy-3-xanthanone, 4′,5′-Bis[bis(2-pyridylmethyl)aminomethyl]-2′,7′-dichlorofluorescein

CDX-Z0001-M020 CDX-Z0001-M125
102991-220EA 301.85 USD
102991-220 102991-222
Zinpyr-1 ≥95%
Zinpyr-1

Membrane permeable fluorophore suitable for the fluorometric detection of Zn2+. In living cells Zinpyr-1 mainly stains the Golgi or a Golgi-associated vesicle.

Formula: C₄₆H₃₆Cl₂N₆O₅
Storage Temperature: Freezer
MDL Number: MFCD04039712
CAS Number: 288574-78-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR