Vindoline ≥98%

Supplier: ADIPOGEN CORP MS
Warning

CDX-V0002-M050 CDX-V0002-M250 CDX-V0002-GG25
102991-160EA 105.23 USD
102991-160 102991-162 102991-164
Vindoline ≥98%
Vindoline

Reference standard.

Formula: C₂₅H₃₂N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD27991347
CAS Number: 2182-14-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR