meso-Tetraphenyl-tetrabenzoporphine palladium complex ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: PdPh4TBP

CDX-T0083-M025 CDX-T0083-M250
102991-096EA 299.08 USD
102991-096 102991-098
meso-Tetraphenyl-tetrabenzoporphine palladium complex ≥97%
meso-Tetraphenyl-tetrabenzoporphine palladium complex

Phosphorescent probe for measuring oxygen in very low concentrations. Luminescent marker for oxygen and pH in biomedical imaging. Used as sensitizer in the process of photon up-conversion based on triplet–triplet annihilation (TTA). Used as agent for PDT, optical limiters and other types of nonlinear optical materials.

Formula: C₆₀H₃₆N₄Pd
Storage Temperature: Refrigerator
MDL Number: MFCD03095471
CAS Number: 119654-64-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (NMR)
Appearance/Color Deep blue powder.
Solubility Soluble in dichloromethane, toluene or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR