Tetramethylrhodamine-6-maleimide ≥97%

Supplier: ADIPOGEN CORP MS
Warning

CDX-T0030-M002 CDX-T0030-M025
102991-056EA 263.08 USD
102991-056 102991-058
Tetramethylrhodamine-6-maleimide ≥97%
Tetramethylrhodamine-6-maleimide

Fluorescent reagent with high selectivity.

Formula: C₂₈H₂₃N₃O₅
Storage Temperature: Refrigerator
CAS Number: 174568-68-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in methanol, DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR