Sulfamethoxazole hydroxylamine (synthetic) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: SMX-HA

CDX-S0056-M010 CDX-S0056-M025
102991-018EA 279.69 USD
102991-018 102991-020
Sulfamethoxazole hydroxylamine (synthetic) ≥98% (by HPLC)
Sulfamethoxazole hydroxylamine

Sulfamethoxazole hydroxylamine is a metabolite of sulfamethoxazole, a sulfonamide bacteriostatic antibiotic.


  • Shown to induce lymphocyte toxicity
  • Suppression of proliferation
  • Analytical reference material


Has been shown to be an important component in sulfonamide hypersensitivity reactions and has been implicated in sulfonamide-mediated immunomodulatory effects. Shown to induce lymphocyte toxicity and suppression of proliferation. Compound can be used as analytical reference material.


Off-white powder


Soluble in methanol or DMSO. Sligthly soluble in water

Formula: C₁₀H₁₁N₃O₄S
Storage Temperature: Freezer
CAS Number: 114438-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR