Rhodamine 110 ≥97%
Supplier: ADIPOGEN CORP MS
Synonyms:
Rhodamine 110 chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid hydrochloride
Fluorescent dye. Fluorophore standard that is widely used in enzyme assay systems. Precursor for a variety of R110-based enzyme substrates for flow cytometric measurements. Used as a calibration standard for R110-based enzyme substrates. Compared to AMC and AFC enzyme substrates, fluorogenic R110 substrates are generally more sensitive, giving longer absorption and fluorescence wavelength than AMC and AFC substrates. Spectral data: lambdaex 498nm| lambdaem 520nm in methanol.
Formula:
C₂₀H₁₅ClN₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00042009 CAS Number: 13558-31-1 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | Red to red-brown crystalline or powder. |
Solubility | Soluble in ethanol, methanol, DMSO or DMF. Slightly soluble in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...