(±)-Potassium citramalate monohydrate ≥97%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (±)-2-Hydroxy-2-methylsuccinic acid dipotassium salt

CDX-P0157-G001 CDX-P0157-G005 CDX-P0157-G025
102990-884EA 96.92 USD
102990-884 102990-886 102990-888
(±)-Potassium citramalate monohydrate ≥97%
(±)-Potassium citramalate monohydrate

Building block for synthesis.

Formula: C₅H₆O₅·2K·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD18252952
CAS Number: 1030365-02-6

Order Now

Specification Test Results

Source/Host Synthetic
Purity 97% (NT)
Appearance/Color White crystalline.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR