4-Isopropylantipyrine

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Isopropyl-2,3-dimethyl-1-phenyl-5-pyrazolone, Propyphenazone, 4-Isopropyl-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one

CDX-P0090-M100 CDX-P0090-G001
102990-814EA 88.62 USD
102990-814 102990-816
4-Isopropylantipyrine
4-Isopropylantipyrine

Analgesic, antipyretic, and anti-inflammatory compound.

Formula: C₁₄H₁₈N₂O
MDL Number: MFCD00053368
CAS Number: 479-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR