4-Methylumbelliferyl oleate ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
4-Methyl-2-oxo-2H-chromen-7-yl 9-octadecenoate
CDX-M0086-M025
CDX-M0086-M250
CDX-M0086-GG25
102990-470EA
84.46
USD
102990-470
102990-472
102990-474
4-Methylumbelliferyl oleate ≥95%
4-Methylumbelliferyl oleate
Fluorogenic substrate for lipases, which can be used in aqueous solutions as well as several organic solvents.
Formula:
C₂₈H₄₀O₄ Storage Temperature: Refrigerator |
CAS Number:
18323-58-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white solidified mass. |
Solubility | Soluble in 2-Methoxyethanol, DMSO or chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...