5-Methoxy-2,3,3-trimethyl-1-(4-sulfobutyl)indolium inner salt ≥95%

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0049-M500 CDX-M0049-GG25
102990-416EA 181.38 USD
102990-416 102990-418
5-Methoxy-2,3,3-trimethyl-1-(4-sulfobutyl)indolium inner salt ≥95%
5-Methoxy-2,3,3-trimethyl-1-(4-sulfobutyl)indolium inner salt

Fluorescent reagent.

Formula: C₁₆H₂₃NO₄S
Storage Temperature: Freezer
CAS Number: 54136-27-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR