8-Hydroxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide ≥95%

Supplier: ADIPOGEN CORP MS
Warning

CDX-H0043-M010 CDX-H0043-M050
102990-236EA 254.77 USD
102990-236 102990-238
8-Hydroxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide ≥95%
8-Hydroxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide

Fluorescent pH indicator for the weakly acidic pH range and enzyme substrate reference standard.

Formula: C₂₂H₂₅N₃O₇S₃
Storage Temperature: Freezer
CAS Number: 127044-59-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO, DMF or alcohols.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR