IPB ≥90%

Supplier: ADIPOGEN CORP MS

Synonyms: 4-[3-(4-Iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulfonate sodium salt, 4-[3-(4-Iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulphonate sodium salt

CDX-I0004-M250 CDX-I0004-G001
102990-144EA 228.46 USD
102990-144 102990-146
IPB ≥90%
IPB

4-[3-(4-iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulfonate sodium salt (IPB) have found application in clinical assays and cell proliferation assays that are based on a spectrophotometric measurement of dehydrogenase activities of cells. Disulfonated tetrazolium salt IPB, that produces a highly water-soluble formazan dye that proved to be extremely useful as a cell-viability indicator. IPB is reduced by reduced nicotinamide adenine dinucleotide (NADH) to formazan in the presence of a redox mediator such as 1-methoxy PMS, which provides the basis to specrophotometric assays of NADH.

Formula: C₁₉H₁₀IN₆NaO₁₀S₂
Storage Temperature: Freezer
CAS Number: 161617-45-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Pale yellow powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR