Halofenozide ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: RG 100864

CDX-H0097-M050 CDX-H0097-M250
102990-074EA 245.08 USD
102990-074 102990-076
Halofenozide ≥98%
Halofenozide

Dibenzoylhydrazine based nonsteroidal insecticide. Ecdysone agonist that mimics the biological function of the natural insect molting hormone 20-hydroxyecdysone, causing premature molting and interfering with the normal growth and development in insects. Ecdysteroid agonist are also used as ligands for gene expression and gene therapy research. Compound can be used as analytical reference material.

Formula: C₁₈H₁₉ClN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD03158909
CAS Number: 112226-61-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR