Sodium galacturonate anhydrous ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: D-(+)-Galacturonic acid sodium salt

CDX-G0007-G005 CDX-G0007-G025 CDX-G0007-G250
102990-050EA 132.92 USD
102990-050 102990-052 102990-054
Sodium galacturonate anhydrous ≥98%
Sodium galacturonate anhydrous

A sugar acid sodium salt preparation.

Formula: C₆H₉NaO₇
Storage Temperature: Refrigerator
MDL Number: MFCD06201013
CAS Number: 14984-39-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (Titration)
Appearance/Color White to off-white crystalline powder.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR