GSH-OEt, AdipoGen Life Sciences
Supplier: ADIPOGEN CORP MS
CDX-G0006-M100 CDX-G0006-M500 CDX-G0006-G005
102990-044EA 96.92 USD
102990-044 102990-046 102990-048
GSH-OEt, AdipoGen Life Sciences
Proteins and Peptides
GSH-OEt is cellpermeable and has been used to protect cells against radiation damage, oxidants and various toxic compounds including heavy metals. GSH-OEt is a protective agent against cellular damage, such as cataracts and mitochondrial degeneration.
It undergoes hydrolysis by intracellular esterases thereby increasing intracellular GSH concentration in many tissues and cell types. Glutathione monoethyl ester may be used to supplement cellular pools of GSH in vitro and in vivo and can prevent ROS formation, neutralize toxic products and block apoptosis pathway. The effect of glutathione (GSH) and glutathione ethyl ester (GSH-OEt) supplementation on GSH homeostasis and exercise-induced oxidative stress was also examined. Glutathione ethyl ester protects against cisplatin-induced ototoxicity in the rat.