Furanone C-56 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

CDX-F0046-M010 CDX-F0046-M100 CDX-F0046-G001
102989-962EA 175.85 USD
102989-962 102989-964 102989-966
Furanone C-56 ≥95%
Furanone C-56

Furanone compounds and lactam analogues.

Formula: C₅H₃BrO₂
Storage Temperature: Freezer
CAS Number: 199744-38-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO, ethanol or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR