6-(Dimethylamino)purine

Supplier: ADIPOGEN CORP MS

CDX-D0318-G001
102989-766EA 175.85 USD
102989-766
6-(Dimethylamino)purine
6-(Dimethylamino)purine

A puromycin analog that acts as a protein kinase inhibitor. Selectively induces premature mitosis in mammalian cells. Inhibits TNF-alpha upregulation of ICAM-1, lipolysis, and the activation of JNK/SAPK. Inhibits p34cdc2/cyclin B (IC50 = 300 µM). Induces apoptosis in S-phase-arrested HeLa cells (~5 mM).

Formula: C₇H₉N₅
MDL Number: MFCD00005573
CAS Number: 938-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR