Collagenase chromophore substrate component A ≥95%

Supplier: ADIPOGEN CORP MS

CDX-C0079-M010 CDX-C0079-M050 CDX-C0079-G001
102989-444EA 96.92 USD
102989-444 102989-446 102989-448
Collagenase chromophore substrate component A ≥95%
Collagenase chromophore substrate component A

Enzyme substrate.

Formula: C₃₈H₅₂N₁₀O₈·2H₂O
Storage Temperature: Freezer
CAS Number: 118081-33-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Orange powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR