N-Biotinyl-NH-(PEG)2-COOH DIPEA ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: O-(N-Biotinyl-3-aminopropyl)-O'-(N-glutaryl-3-aminopropyl)-diethyleneglycol . DIPEA

CDX-B0209-M050 CDX-B0209-M250
102989-400EA 99.69 USD
102989-400 102989-402
N-Biotinyl-NH-(PEG)2-COOH DIPEA ≥95%
N-Biotinyl-NH-(PEG)2-COOH DIPEA

Functionalized biotin. Biotinylated polyethylene glycol (PEG-Biotin) is an avidine or streptavidin reactive PEG derivative that can be used to modify biomolecues or other surfaces through biotin-streptavidin interaction. Used for addition of biotin-PEG to the N-terminus of resin-bound peptides. Can be introduced using standard coupling methods, such as PyBOP or TBTU and has better solubility in DMF and NMP than comparable non-PEG-containing derivatives, such as biotin-Ahx and biocytin.

Formula: C₂₅H₄₄N₄O₈S·C₈H₁₉N
Storage Temperature: Freezer
CAS Number: 1205744-09-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR