AMCA-X-N-succinimidyl ester ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
AMCA-X, SE, 6-((7-Amino-4-methylcoumarin-3-acetyl)amino) hexanoic acid, N-succinimidyl ester
CDX-A0074-M010
CDX-A0074-M100
102989-184EA
318.46
USD
102989-184
102989-186
AMCA-X-N-succinimidyl ester ≥95%
AMCA-X-N-succinimidyl ester
AMCA-X SE is an amine-reactive, UV-excitable, blue fluorescent dye.
Formula:
C₂₂H₂₅N₃O₇ MW: 443.45 g/mol Storage Temperature: Freezer |
CAS Number:
216309-02-3 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Light yellow powder. |
Solubility | Soluble in DMSO or DMF. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...