AMCA-X ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
6-((7-Amino-4-methylcoumarin-3-acetyl)amino) hexanoic acid
CDX-A0073-M050
CDX-A0073-M250
102989-180EA
376.62
USD
102989-180
102989-182
AMCA-X ≥95%
AMCA-X
Amine-reactive blue fluorescent probe.
Formula:
C₁₈H₂₂N₂O₅ Storage Temperature: Freezer |
CAS Number:
205124-69-2 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or acetonitrile. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...