Cochliodone A ≥98%

Supplier: ADIPOGEN CORP MS
Warning

BVT-0458-M001
102989-084EA 207.69 USD
102989-084
Cochliodone A ≥98%
Cochliodone A

Dimeric spiro-azaphilone. Mycotoxin.

Formula: C₃₄H₃₈O₁₂
Storage Temperature: Refrigerator
CAS Number: 1072931-48-6

Order Now

Specification Test Results

Source/Host Isolated from Chaetomium sp.
Purity 98% (HPLC, NMR)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO, acetone, dichloromethane or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR