Rugulosin (from Penicillium species) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0444-M005
102989-040EA 456.92 USD
102989-040
Rugulosin (from Penicillium species) ≥98% (by HPLC)
Rugulosin

Rugulosin is an anti-viral, HIV-1 integrase inhibitor.


  • Inhibitor of N-Hsp90
  • Antibiotic
  • Mycotoxin


Antibiotic (Streptococcus, Corynebacterium, MRSA). DNA replication, transcription and repair inhibitor (carcinogenic activity). RNA polymerase and ribonuclease H inhibitor. Insecticidal activity.


Yellow solid


Soluble in DMSO, acetone, ethanol or methanol

Storage Temperature: Freezer
CAS Number: 23537-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR