Averantin ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: BRN 2309929

BVT-0169-M001 BVT-0169-M005
102988-898EA 110.77 USD
102988-898 102988-900
Averantin ≥98%
Averantin

Averantin is a cytotoxic against human solid tumor cell lines.


  • Mycotoxin
  • Cytotoxic
  • Intermediate to Aflatoxin B1


Intermediate of the biosynthetic pathway to Aflatoxin B1. Cytotoxic against human solid tumor cell lines.


Orange powder


Soluble in acetone, methanol or DMSO

Formula: C₂₀H₂₀O₇
Storage Temperature: Freezer
CAS Number: 5803-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR