Kojic acid

Supplier: ADIPOGEN CORP MS

Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone

BVT-0019-M010
102988-874EA 62.31 USD
102988-874
Kojic acid
Kojic acid

Mycotoxin. Chelation agent with polyfunctional skeleton for development of biologically active compounds. Moderate antibacterial agent. Tyrosinase inhibitor. Inhibits catecholase activity of tyrosinase. Anti-melanogenic. Skin whitening agent. Antioxidant. Reactive oxygen species (ROS) scavenger. D-Amino acid oxidase (DAAO) inhibitor. Shown to inhibit NF-kB activation. Inducer of macrophage activation. Radioprotectant activity. Antileishmanial agent. Insecticidal. Antifungal chemosensitizer. Used in cosmetics and as food additive.

Formula: C₆H₆O₄
MDL Number: MFCD00006580
CAS Number: 501-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR