6-Amino-8-trifluoromethylphenanthridine ≥99%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
6A-8tFP
AG-MR-C0031-M001
AG-MR-C0031-M005
AG-MR-C0031-M025
102988-864EA
83.08
USD
102988-864
102988-866
102988-868
6-Amino-8-trifluoromethylphenanthridine ≥99%
6-Amino-8-trifluoromethylphenanthridine
Potent antiprion agent. Higher activity than 6-Aminophenanthridine . Ribosome-borne protein folding activity (RPFA) inhibitor. Binds to the ribosomal RNA and inhibits specifically the protein folding activity of the ribosome. Inhibitor of protein aggregation.
Formula:
C₁₄H₉F₃N₂ Storage Temperature: Freezer |
CAS Number:
651055-83-3 |
Specification Test Results
Purity |
|
Appearance/Color | Beige solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...