(R)-Perharidine 1 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

AG-MR-C0013-M001 AG-MR-C0013-M005 AG-MR-C0013-M025
102988-824EA 96.92 USD
102988-824 102988-826 102988-828
(R)-Perharidine 1 ≥95%
(R)-Perharidine 1

Potent and selective inhibitor of cyclin dependent kinases CDK1, CDK2, CDK5 and CDK9. Apoptosis inducer. Inhibits the proliferation of various cancer cell lines. Casein kinase 1 (CK1delta/epsilon) inhibitor. Potential anti-inflammatory compound. Potential antidiabetic compound.

Formula: C₂₅H₃₀N₆O
Storage Temperature: Freezer
CAS Number: 1133437-80-5

Order Now

Specification Test Results

Purity 95% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR