α-Galactosylceramide analog 8 ≥96%

Supplier: ADIPOGEN CORP MS

Synonyms: α-Gal-Cer Analog 8

AG-CR1-3622-C500
102988-804EA 332.31 USD
102988-804
α-Galactosylceramide analog 8 ≥96%
alpha-Galactosylceramide analog 8

Immunostimulant. Potent stimulator of natural killer T (NKT) cells. Specific ligand of the lipid-binding MHC class I-like protein CD1d in human and mouse NKT cells. Potent antitumor compound. Stimulates the production of smaller amounts of IFN-gamma than alpha-GalCer, but induces equal or greater IL-4 secretion in mouse models.

Formula: C₅₀H₉₇N₃O₈
Storage Temperature: Freezer
CAS Number: 922727-14-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 96% (NMR)
Appearance/Color White solid.
Solubility Soluble in pyridine, methanol, 100% ethanol, DMSO or dimethylformamide.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR