Tripolin B ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: (3E)-1,3-Dihydro-3-(1H-imidazol-5-ylmethylene)-2H-indol-2-one

AG-CR1-3521-M001
102988-578EA 131.54 USD
102988-578
Tripolin B ≥98%
Tripolin B

Aurora A and B modulator.

Formula: C₁₂H₉N₃O
Storage Temperature: Freezer
CAS Number: 372164-71-1

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Yellow solid.
Solubility Soluble in DMSO (at least 5 mg/ml).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR