Necrostatin-1
Supplier: ADIPOGEN CORP MS
Synonyms:
5-((1H-Indol-3-yl)methyl)-3-methyl-2-thioxoimidazolidin-4-one, Nec-1
Competitive indoleamine 2,3-dioxygenase (IDO) inhibitor. Cell permeable, potent and selective necroptosis (a non-apoptotic form of programmed cell death) inhibitor. Showed neuroprotection in a murine model of ischemic brain injury in vivo. Selective and ATP-competitive receptor-interacting protein kinase 1 (RIPK1) inhibitor. Used in inflammatory and degenerative disease models to target RIPK1. Suppresses autophagy and apoptosis in murine brain injury models. Cardioprotective and neuroprotective.
Formula:
C₁₃H₁₃N₃OS MW: 259.327 g/mol |
MDL Number:
MFCD00056916 CAS Number: 4311-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...