Synthetic Antipain Dihydrochloride (from Synthetic)
Supplier: ADIPOGEN CORP MS
Potent reversible inhibitor of serine/cysteine proteases and some trypsin-like proteases. Inhibits papain, trypsin and trypsin-like serine proteases as well as cathepsin A, B and D. Similar activity spectrum comparable to leupeptin (AG-CP3-7000). Acts by forming a hemiacetal adduct between its aldehyde group and the active serine of the proteinase. Inhibits transformation of NIH3T3 cells after transfection with an activated H-ras oncogene. Used to evaluate the role of proteases in the process of malignant cell transformations. Shows antibacterial and antiparasitic activity.
- White to off-white solid
- Soluble in water, ethanol, methanol, DMSO or acetonitrile