Artesunate

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Oxo-4-(((3R,5aS,6R,8aS,9R,10R,12R,12aR)-3,6,9-trimethyldecahydro-3H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10-yl)oxy)butanoic acid

AG-CN2-0469-M100 AG-CN2-0469-M500 AG-CN2-0469-G001
102988-386EA 69.23 USD
102988-386 102988-388 102988-390
Artesunate
Artesunate

Semi-synthetic derivative of artemisinin. Anti-malaria. Anti-schistosomiasis. Inhibitor of human cytomegalovirus (CMV). Apoptosis inducer. Anti-cancer. Immunosuppressant. Inhibits production of tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6), monocyte chemotactic protein-1 (MCP-1) and nitric oxide (NO).

Formula: C₁₉H₂₈O₈
MDL Number: MFCD00866204
CAS Number: 88495-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR