(+)-Rutin trihydrate ≥95%

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0408-G005
102988-162EA 34.62 USD
102988-162
(+)-Rutin trihydrate ≥95%
(+)-Rutin trihydrate

Antioxidant. Free radical scavenger. Nitric oxide (NO) scavenger. Anticancer compound. Antiproliferative. Apoptosis inducer. Reduces induced DNA damage. Protective against carcinogenesis. Hypolipidaemic. Reduces triacylglycerol levels. Anti-inflammatory. Platelet aggregation inhibitor. Anti-hyperglycaemic. Anti-adipogenic. Insulin-mimetic. Activates synthesis of glucose transporter GLUT4. Neuroprotective. Cardioprotective. Reduces lipid peroxidation. Weak alpha-glucosidase inhibitor. Vasodilatory compound.

Formula: C₂₇H₃₀O₁₆·3H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00149490
CAS Number: 250249-75-3

Order Now

Specification Test Results

Source/Host Isolated from Ruta graveolens.
Purity 95% (NMR)
Appearance/Color Pale yellow powder.
Solubility Soluble in DMSO, acetone, dichloromethane or ethyl acetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR