Asiatic acid

Supplier: ADIPOGEN CORP MS

AG-CN2-0400-M005 AG-CN2-0400-M025
102988-146EA 55.38 USD
102988-146 102988-148
Asiatic acid
Asiatic acid

Apoptosis inducer. Cell cycle arrest inducer. Anticancer compound Antioxidant. Hepatoprotective. Inhibits TGF-beta/Smad-mediated fibrogenesis. Stimulates wound healing. Anti-diabetic. Glycogen phosphorylase inhibitor. Neuroprotective. Modulates multiple targets associated with amyloid-beta precursor protein processing and amyloid-beta protein clearance. Down regulates BACE1 and increases ADAM10 maturation. Anti-hyperglycemic compound. Anti-inflammatory and antinociceptive compound. Antiangiogenic. Acetylcholinesterase (AChE) inhibitor. PPARgamma inhibitor through a C/EBPbeta-independent mechanisms. Anti-osteoporotic. Inhibits adipogenic differentiation of bone marrow stromal cells (BMSC).

Formula: C₃₀H₄₈O₅
MDL Number: MFCD00238541
CAS Number: 464-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR