Pyranonigrin A ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (7R)-6,7-Dihydro-3,7-dihydroxy-2-[(E)-1-propene-1-yl]-pyrano[2,3-c]pyrrole-4,5-dione

AG-CN2-0156-C250 AG-CN2-0156-M001
102988-106EA 103.85 USD
102988-106 102988-108
Pyranonigrin A ≥95%
Pyranonigrin A

Antioxidant. Radical scavenging activity towards DPPH and superoxide. Shows a suppressive effect on the expression of vascular cell adhesion molecule-1 (VCAM-1) in human umbilical vein endothelial cells (HUVECs) induced by tumor necrosis factor-alpha (TNF-alpha). Insecticidal agent.

Formula: C₁₀H₉NO₅
Storage Temperature: Freezer
CAS Number: 773855-65-5

Order Now

Specification Test Results

Source/Host Isolated from Aspergillus niger FKI-2633.
Purity 95% (HPLC)
Appearance/Color Crystalline solid.
Solubility Soluble in DMSO or hot methanol. Insoluble in hexane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR