Calpinactam ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: FKI-4905

AG-CN2-0131-C250 AG-CN2-0131-M001
102988-018EA 138.46 USD
102988-018 102988-020
Calpinactam ≥95%
Calpinactam

Specific anti-mycobacterial. Inhibits growth of Mycobacteria smegmatis but also Mycobacteria tuberculosis. M. smegmatis is used as a model for mycobacterial species. It is useful for the research analysis of other Mycobacteria species in laboratory experiments. M. smegmatis is commonly used in work on the mycobacterium species, since it is a "fast grower" and non-pathogenic.

Formula: C₃₈H₅₇N₉O₈
Storage Temperature: Freezer
CAS Number: 1205538-83-5

Order Now

Specification Test Results

Source/Host Synthetic. Originally isolated from Mortierella alpina.
Purity 95% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR