Fluorescein-5-thiosemicarbazide ≥97% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: 5-FTSC

CDX-F0005-M050 CDX-F0005-M100 CDX-F0005-M500
102987-928EA 166.15 USD
102987-928 102987-930 102987-932
Fluorescein-5-thiosemicarbazide ≥97% (by NMR)
Fluorescein-5-thiosemicarbazide

Labeling of cell-surface functional groups (glycophorins). Detection of protein carbonyls in aging tissues.

Formula: C₂₁H₁₅N₃O₅S
Storage Temperature: Freezer
MDL Number: MFCD04221206
CAS Number: 76863-28-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >97% (NMR)
Appearance/Color Crystalline.
Solubility Soluble in water or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR