Fluorescein-5-thiosemicarbazide ≥97% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
5-FTSC
CDX-F0005-M050
CDX-F0005-M100
CDX-F0005-M500
102987-928EA
166.15
USD
102987-928
102987-930
102987-932
Fluorescein-5-thiosemicarbazide ≥97% (by NMR)
Fluorescein-5-thiosemicarbazide
Labeling of cell-surface functional groups (glycophorins). Detection of protein carbonyls in aging tissues.
Formula:
C₂₁H₁₅N₃O₅S Storage Temperature: Freezer |
MDL Number:
MFCD04221206 CAS Number: 76863-28-0 |
Specification Test Results
Source/Host | Synthetic. |
Purity | >97% (NMR) |
Appearance/Color | Crystalline. |
Solubility | Soluble in water or DMF. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...