3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]

Supplier: ADIPOGEN CORP MS

Synonyms: DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide

CDX-D0130-M100 CDX-D0130-G001
102987-884EA 253.38 USD
102987-884 102987-886
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]

3,3'-Dipropylthiadicarbocyanine iodide is applied for transmembrane potential measurements.

Formula: C₂₅H₂₇IN₂S₂
MDL Number: MFCD00075033
CAS Number: 53213-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR