Ponasterone A

Supplier: ADIPOGEN CORP MS

AG-CN2-0053-M001 AG-CN2-0053-M005 AG-CN2-0053-M025
102987-858EA 83.08 USD
102987-858 102987-860 102987-862
Ponasterone A
Ponasterone A

A member of the ecdysteroid family. Ecdysone receptor (EcR) agonist. Analog of ecdysone with similar properties to muristerone A. Functional, reliable and economical substitute for muristerone A as an inducer for the ecdysone-inducible mammalian expression system. Induces expression of beta-galactosidase to levels similar to those obtained with muristerone A induction. Potent compound for ecdysteroid activity. Insect steroid hormone involved in regulating metamorphosis.

Formula: C₂₇H₄₄O₆
CAS Number: 13408-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR