Sauchinone ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 1′-Epimer dihydrocarpanone, (5aR,5a1R,7R,8S,8aR,14aS)-7,8-Dimethyl-5a,5a1,6,7,8,8a-hexahydro-5H-benzo[kl]bis([1,3]dioxolo)[4,5-b:4',5'-g]xanthen-5-one

AG-CN2-0032-M001 AG-CN2-0032-M005
102987-846EA 41.54 USD
102987-846 102987-848
Sauchinone ≥98%
Sauchinone

Hepatoprotective antioxidant. Anti-inflammatory. Suppresses NF-kB p65 activity. Neuroprotective. Apoptosis inhibitor. Bone resorption/osteoclastogenesis inhibitor. Reduces ROS formation. Antiseptic. AMP-activated kinase (AMPK) activator. Akt phosphorylation inhibitor.

Formula: C₂₀H₂₀O₆
Storage Temperature: Freezer
MDL Number: MFCD08702698
CAS Number: 177931-17-8

Order Now

Specification Test Results

Source/Host Isolated from Saururus chinensis.
Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in DMSO or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR