FAMC ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 3-(4,6-Difluorotriazinyl)amino-7-methoxycoumarin

CDX-D0099-M010 CDX-D0099-M050
102987-796EA 146.77 USD
102987-796 102987-798
FAMC ≥95%
FAMC

Polarity fluorescent probe.

Formula: C₁₃H₈F₂N₄O₃
Storage Temperature: Freezer
CAS Number: 127192-67-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (NMR)
Appearance/Color Solid.
Solubility Soluble in DMSO or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR