7-Diethylaminocoumarin-3,4-dicarboxylic acid ≥95%

Supplier: ADIPOGEN CORP MS

CDX-D0039-M100 CDX-D0039-G001
102987-698EA 278.31 USD
102987-698 102987-700
7-Diethylaminocoumarin-3,4-dicarboxylic acid ≥95%
7-Diethylaminocoumarin-3,4-dicarboxylic acid

Proteomic research material.

Formula: C₁₅H₁₅NO₆
Storage Temperature: Refrigerator
CAS Number: 75240-77-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR