(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: (4aS,7S,8aR)-8,8-Dichloro-9,9-dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide

CDX-D0031-G005 CDX-D0031-G025
102987-672EA 263.08 USD
102987-672 102987-674
(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine ≥97%
(+)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine

Chiral oxidizing agent. Used for asymmetric synthesis as chiral catalyst. Hydroxylation reagent.

Formula: C₁₀H₁₃Cl₂NO₃S
Storage Temperature: Freezer
MDL Number: MFCD00243015
CAS Number: 127184-05-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (Titration)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR