5-Carboxytetramethylrhodamine (5-TAMRA) ≥97%
Supplier: ADIPOGEN CORP MS
Synonyms:
5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer
CDX-C0055-M025
CDX-C0055-M250
102987-638EA
210.46
USD
102987-638
102987-640
5-Carboxytetramethylrhodamine (5-TAMRA) ≥97%
5-Carboxytetramethylrhodamine (5-TAMRA)
Free acid form of 5-carboxytetramethylrhodamine single isomer. Widely used fluorophore for preparing bioconjugates, especially fluorescent antibody and avidin derivatives used in immunochemistry. Used as a dye for oligonucleotide labeling and automated DNA sequencing applications.
Formula:
C₂₅H₂₂N₂O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00269769 CAS Number: 91809-66-4 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in methanol, DMSO or DMF. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...