Colchicine

Supplier: ADIPOGEN CORP MS

CDX-C0032-M100 CDX-C0032-G005 CDX-C0032-G050
102987-584EA 58.15 USD
102987-584 102987-586 102987-588
Colchicine
Colchicine

Binds specifically with tubulin thus interfering with microtubule organization.

Formula: C₂₂H₂₅NO₆
MDL Number: MFCD00078484
CAS Number: 64-86-8
UN: 1544
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR